Learn More
Cayman Chemical 3'-O-Methylguanosine, Purity: ≥98%, CAS: 10300-27-3, 100 mg
SDP
Catalog No. 502943528
Encompass_Preferred
An antiviral nucleoside analog; inhibits viral plaque formation and viral RNA synthesis in vaccinia virus-infected Vero cells at 10 µM
Catalog No. 50-294-3528 Supplier Cayman Chemical Supplier No. 40292100 MG
An antiviral nucleoside analog; inhibits viral plaque formation and viral RNA synthesis in vaccinia virus-infected Vero cells at 10 µM