Learn More
Cayman Chemical 3-O-MethylgunosIn 50mg
SDP
Catalog No. 502944834
Encompass_Preferred
An antiviral nucleoside analog; inhibits viral plaque formation and viral RNA synthesis in vaccinia virus-infected Vero cells at 10 µM
Catalog No. 50-294-4834 Supplier Cayman Chemical Supplier No. 4029250 MG
An antiviral nucleoside analog; inhibits viral plaque formation and viral RNA synthesis in vaccinia virus-infected Vero cells at 10 µM