Learn More
Medchemexpress LLC 3-O-Methylquercetin | 1486-70-0 | 316.26 | 5 MG
SDP
Catalog No. 5000414678
Encompass_Preferred
3-O-Methylquercetin acts as an inhibitor of cAMP and cGMP-phosphodiesterase (PDE) with IC50 values of 13.8 μM and 14.3 μM, respectively. It also inhibits β-secretase with an IC50 of 6.5 μM. This compound demonstrates neuroprotective effects against neuronal death caused by oxidative damage and exhibits strong antiviral activity against poliovirus, coxsackie virus, and human rhinovirus. Additionally, it has anti-inflammatory and trachea-relaxing properties, making it relevant for the study of inflammatory diseases and asthma.Key characteristics include:
  • Inhibits cAMP and cGMP-phosphodiesterase.
  • Inhibits β-secretase.
  • Provides neuroprotection against oxidative damage.
  • Exhibits strong antiviral activity.
  • Possesses anti-inflammatory and trachea-relaxing effects.
Catalog No. 50-004-14678 Supplier Medchemexpress LLC Supplier No. HYN18605MG
3-O-Methylquercetin acts as an inhibitor of cAMP and cGMP-phosphodiesterase (PDE) with IC50 values of 13.8 μM and 14.3 μM, respectively. It also inhibits β-secretase with an IC50 of 6.5 μM. This compound demonstrates neuroprotective effects against neuronal death caused by oxidative damage and exhibits strong antiviral activity against poliovirus, coxsackie virus, and human rhinovirus. Additionally, it has anti-inflammatory and trachea-relaxing properties, making it relevant for the study of inflammatory diseases and asthma.Key characteristics include:
  • Inhibits cAMP and cGMP-phosphodiesterase.
  • Inhibits β-secretase.
  • Provides neuroprotection against oxidative damage.
  • Exhibits strong antiviral activity.
  • Possesses anti-inflammatory and trachea-relaxing effects.