Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Octen-2-one 96.0+%, TCI America™
SDP

Catalog No. O025225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O025225ML 25 mL
O02525ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O025225ML Supplier TCI America Supplier No. O025225ML
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1669-44-9
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00015565
InChI Key ZCFOBLITZWHNNC-VOTSOKGWSA-N
Synonym 3-octen-2-one, trans-3-octen-2-one, e-3-octen-2-one, 3e-3-octen-2-one, e-oct-3-en-2-one, unii-m26ah283xv, 3-octen-2-one, e, 3e-octen-2-one, 3e-oct-3-en-2-one, hex-1-enyl methyl ketone
PubChem CID 5363229
IUPAC Name (E)-oct-3-en-2-one
SMILES CCCCC=CC(=O)C

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Octen-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.