Learn More
Apexbio Technology LLC 3-OH-4-OME CINNAMIC ACID-20mg
SDP
Catalog No. 5000897371
Encompass_Preferred
3-Hydroxy-4-methoxycinnamic acid (CAS No 537-73-5) is a compound in biomedical research primarily focusing on its antioxidant and anti-inflammatory properties It originates from natural sources like plants where it serves as a secondary metabolite The mechanism of action typically involves modulation of oxidative stress pathways particularly targeting reactive oxygen species and related signaling cascades In vitro it shows activity in the nanomolar to low micromolar range indicating its potency in biological assays Its applications range from studies involving cell models such as fibroblasts and epithelial cells to animal studies where it is used to investigate protective effects against oxidative damage and inflammatory conditions It is also employed in drug screening for its potential therapeutic value Experimental concentrations typically depend on the specific research design ensuring that outcomes are aligned with study objectives
Catalog No. 50-008-97371 Supplier Apexbio Technology LLC Supplier No. N139320
3-Hydroxy-4-methoxycinnamic acid (CAS No 537-73-5) is a compound in biomedical research primarily focusing on its antioxidant and anti-inflammatory properties It originates from natural sources like plants where it serves as a secondary metabolite The mechanism of action typically involves modulation of oxidative stress pathways particularly targeting reactive oxygen species and related signaling cascades In vitro it shows activity in the nanomolar to low micromolar range indicating its potency in biological assays Its applications range from studies involving cell models such as fibroblasts and epithelial cells to animal studies where it is used to investigate protective effects against oxidative damage and inflammatory conditions It is also employed in drug screening for its potential therapeutic value Experimental concentrations typically depend on the specific research design ensuring that outcomes are aligned with study objectives