Learn More
Medchemexpress LLC 3'OMe-m7GpppAmpG | 2089461-55-0 | C33H45N15O24P4 | 11.6 MG
SDP
Catalog No. 5000354249
Encompass_Preferred
3'OMe-m7GpppAmpG is a trinucleotide Cap analogue that demonstrates significant translational efficiency. It is a potential molecular biology tool for mRNA vaccines and mRNA transfection for applications such as protein production, gene therapy, and anti-cancer immunization. The free form is prone to instability, so a stable salt form (3'OMe-m7GpppAmpG ammonium solution (100mM)) is advisable for equivalent biological activity.
  • Significant translational efficiency
  • Potential molecular biology tool for mRNA vaccines and mRNA transfection
  • Useful for protein production
  • Applicable in gene therapy
  • Used in anti-cancer immunization
Catalog No. 50-003-54249 Supplier Medchemexpress LLC Supplier No. HY145973A11.6MG
3'OMe-m7GpppAmpG is a trinucleotide Cap analogue that demonstrates significant translational efficiency. It is a potential molecular biology tool for mRNA vaccines and mRNA transfection for applications such as protein production, gene therapy, and anti-cancer immunization. The free form is prone to instability, so a stable salt form (3'OMe-m7GpppAmpG ammonium solution (100mM)) is advisable for equivalent biological activity.
  • Significant translational efficiency
  • Potential molecular biology tool for mRNA vaccines and mRNA transfection
  • Useful for protein production
  • Applicable in gene therapy
  • Used in anti-cancer immunization