Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Oxabicyclo[3.1.0]hexane-2,4-dione

Catalog No. 3911741G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-117-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-117-41G Supplier Sigma Aldrich Supplier No. 3911741G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Oxabicyclo[3.1.0]hexane-2,4-dione

Specifications

CAS 5617-74-3
Melting Point 59°C to 61°C
Boiling Point 100°C to 102°C (5 mmHg)
Molecular Formula C5H4O3
Linear Formula C5H4O3
MDL Number MFCD00126929
Quantity 1 g
Synonym 1,2-Cyclopropanedicarboxylic anhydride
Molecular Weight (g/mol) 112.08
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.