Learn More
Medchemexpress LLC 3-Oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, (4s)- | 102308-32-7 | 98.6% | C11H19NO4 | 10 G
SDP
Catalog No. 5000434984
Encompass_Preferred
(S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is a chiral compound commonly used as a chiral intermediate in asymmetric synthesis and has important applications in organic synthesis. It is a polar aldehyde substrate that can enhance the polarity, solubility, and the possibility of forming hydrogen bonds of compounds.
  • It is a chiral compound.
  • It is used as a chiral intermediate in asymmetric synthesis.
  • It has important applications in the field of organic synthesis.
  • Enhances polarity, solubility, and hydrogen bond formation.
Catalog No. 50-004-34984 Supplier Medchemexpress LLC Supplier No. HY6934810G
(S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is a chiral compound commonly used as a chiral intermediate in asymmetric synthesis and has important applications in organic synthesis. It is a polar aldehyde substrate that can enhance the polarity, solubility, and the possibility of forming hydrogen bonds of compounds.
  • It is a chiral compound.
  • It is used as a chiral intermediate in asymmetric synthesis.
  • It has important applications in the field of organic synthesis.
  • Enhances polarity, solubility, and hydrogen bond formation.