Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Oxetanemethanol | 6246-06-6 | 88.11 | 100 MG
SDP

Catalog No. 5000448541
Encompass_Preferred

3-Oxetanemethanol is a biochemical reagent suitable for life science research. It can be used as a biological material or an organic compound. This product is for research use only.

  • Biochemical reagent for life science research
  • Suitable as a biological material or organic compound
  • High purity of 99.7%
  • Molecular weight: 88.11
  • Chemical formula: C4H8O2
  • Appears as a colorless to light yellow liquid
  • Stable for 3 years at -20°C (pure form)
  • Boiling point: 204.4°C (760 mmHg)
  • Density: 1.093 g/cm3

Catalog No. 50-004-48541 Supplier Medchemexpress LLC Supplier No. HYW004925100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Oxetanemethanol is a biochemical reagent suitable for life science research. It can be used as a biological material or an organic compound. This product is for research use only.

  • Biochemical reagent for life science research
  • Suitable as a biological material or organic compound
  • High purity of 99.7%
  • Molecular weight: 88.11
  • Chemical formula: C4H8O2
  • Appears as a colorless to light yellow liquid
  • Stable for 3 years at -20°C (pure form)
  • Boiling point: 204.4°C (760 mmHg)
  • Density: 1.093 g/cm3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.