Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Oxetanone

Catalog No. 7315362G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-153-62G 2 g
73-153-610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 73-153-62G Supplier Sigma Aldrich Supplier No. 7315362G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Product may polymerize over time during storage.3-oxetanone can be used to synthesize:
  • Various oxetane-containing lead compounds with improved solubility, reduced lipophilicity and amphiphilicity
  • (hydroxymethyl)oxazoles and (hydroxymethyl)thiazoles via single-step microwave mediated reaction with primary amides and thioamides, respectively
  • Oxetane containing spirocycles through thermal 1,3-dipolar cycloaddition reaction with α-amino acids or secondary α-amino acid esters

Specifications

CAS 6704-31-0
Density 1.124 g/mL (at 25°C)
Molecular Formula C3H4O2
Refractive Index n20/D 1.426
Linear Formula C3H4O2
MDL Number MFCD09263255
Quantity 2 g
Synonym 1,3-Epoxy-2-propanone
Molecular Weight (g/mol) 72.06
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.