Learn More
Medchemexpress LLC 3-oxo-3-[[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[5-hydroxy-2-(4-hydroxyphenyl)-4-oxochromen-7-yl]oxyox | 86546-87-4 | 97.0% | 518.42 g·mol⁻¹ | C24H22O13 | 10 MG
SDP
Catalog No. 5000289621
Encompass_Preferred
Apigenin 7-O-malonylglucoside is a flavonoid glycoside found in chrysanthemum flowers, supplied for research use as a high-purity analytical standard. It is used in studies of natural product chemistry, flavonoid metabolism, and bioactivity assays, and as a reference compound for chromatographic and mass-spectrometric method development.
  • Natural product flavonoid glycoside found in chrysanthemum flowers.
  • Useful as a reference standard for HPLC and LC-MS method development.
  • Suitable for in vitro studies of flavonoid metabolism and bioactivity.
  • High purity (>97%) appropriate for analytical and biochemical research.
  • Provided as a solid in milligram-scale quantities for laboratory use.
Catalog No. 50-002-89621 Supplier Medchemexpress LLC Supplier No. HYN249610MG
Apigenin 7-O-malonylglucoside is a flavonoid glycoside found in chrysanthemum flowers, supplied for research use as a high-purity analytical standard. It is used in studies of natural product chemistry, flavonoid metabolism, and bioactivity assays, and as a reference compound for chromatographic and mass-spectrometric method development.
  • Natural product flavonoid glycoside found in chrysanthemum flowers.
  • Useful as a reference standard for HPLC and LC-MS method development.
  • Suitable for in vitro studies of flavonoid metabolism and bioactivity.
  • High purity (>97%) appropriate for analytical and biochemical research.
  • Provided as a solid in milligram-scale quantities for laboratory use.