Learn More
AdipoGen 3-oxo-C10-L-HSL
SDP
Catalog No. 502560306
Encompass_Preferred
Chemical. CAS 147795-40-2. Formula C14H23NO4. MW 269.3. Synthetic. N-3-Oxodecanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. It was used as an autoinducer of quorum signaling by Pseudomonas putida, Yersinia enterocolitica and other Gram-negative bacteria.
Catalog No. 50-256-0306 Supplier AdipoGen Supplier No. CDXO0059M100
Chemical. CAS 147795-40-2. Formula C14H23NO4. MW 269.3. Synthetic. N-3-Oxodecanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. It was used as an autoinducer of quorum signaling by Pseudomonas putida, Yersinia enterocolitica and other Gram-negative bacteria.