Learn More
AdipoGen 3-oxo-C12-L-HSL
SDP
Supplier:  AdipoGen CDXO0031M020
Encompass_Preferred
Chemical. CAS 168982-69-2. Formula C16H27NO4. MW 297.39. Synthetic. Quorum sensing agent.
Catalog No. 50-256-0342