Learn More
AdipoGen 3-oxo-C14-DL-HSL
SDP
Catalog No. 502560314
Encompass_Preferred
Chemical. CAS 503610-29-5. Formula C18H31NO4. MW 325.4. Synthetic. N-3-Oxotetradecanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. It appears later than shorter acyl chain AHLs in developing biofilms and like other long chain 3-oxo-AHLs, stimulates the production of putisolvin, which in turn inhibits biofilm formation.
Catalog No. 50-256-0314 Supplier AdipoGen Supplier No. CDXO0055M010
Chemical. CAS 503610-29-5. Formula C18H31NO4. MW 325.4. Synthetic. N-3-Oxotetradecanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. It appears later than shorter acyl chain AHLs in developing biofilms and like other long chain 3-oxo-AHLs, stimulates the production of putisolvin, which in turn inhibits biofilm formation.