Learn More
AdipoGen 3-oxo-C16-DL-HSL
SDP
Catalog No. 502560312
Encompass_Preferred
Chemical. CAS 1071479-82-7. Formula C20H35NO4. MW 353.5. Synthetic. N-3-Oxohexadecanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-256-0312 Supplier AdipoGen Supplier No. CDXO0056M010
Chemical. CAS 1071479-82-7. Formula C20H35NO4. MW 353.5. Synthetic. N-3-Oxohexadecanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.