Learn More
AdipoGen 3-oxo-C16-L-HSL
SDP
Supplier:  AdipoGen CDXO0061M010
Encompass_Preferred
Chemical. CAS 925448-37-9. Formula C20H35NO4. MW 353.5. Synthetic. N-3-Oxohexadecanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-256-0303