Learn More
AdipoGen N-(3-Oxopentanoyl)-L-homoserine lactone (10 mg)
SDP
Supplier:  AdipoGen CDXO0139M010
Encompass_Preferred
Chemical. CAS: 148433-21-0. Formula: C9H13NO4. MW: 199.2. Synthetic. N-(3-Oxopentanoyl)-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. Numerous species of bacteria employ 3OC5-HSL in cell-to-cell communication.
Catalog No. 50-205-4578