Learn More
AdipoGen 3-oxo-C8-DL-HSL
SDP
Catalog No. 502560320
Encompass_Preferred
Chemical. CAS 106983-27-1. Formula C12H19NO4. MW 241.3. Synthetic. N-3-Oxooctanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. This compound stimulates the tra gene expression. It is an autoinducer and potent antagonist.
Catalog No. 50-256-0320 Supplier AdipoGen Supplier No. CDXO0052M025
Chemical. CAS 106983-27-1. Formula C12H19NO4. MW 241.3. Synthetic. N-3-Oxooctanoyl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. This compound stimulates the tra gene expression. It is an autoinducer and potent antagonist.