Learn More
AdipoGen 3-Oxo-C9-L-HSL
SDP
Catalog No. 502560295
Encompass_Preferred
Chemical. CAS 441350-70-5. Formula C13H21NO4. MW 255.31. N-3-Oxononanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-256-0295 Supplier AdipoGen Supplier No. CDXO0153M050
Chemical. CAS 441350-70-5. Formula C13H21NO4. MW 255.31. N-3-Oxononanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.