Learn More
Cayman Chemical 3-oxo DeoxycholIc AcId 10mg
SDP
Catalog No. 502913405
Encompass_Preferred
A secondary bile acid; a metabolite of deoxycholic acid in C. perfringens
Catalog No. 50-291-3405 Supplier Cayman Chemical Supplier No. 3522710 MG
A secondary bile acid; a metabolite of deoxycholic acid in C. perfringens