Learn More
Cayman Chemical 3-oxo DeoxycholIc AcId 25mg
SDP
Catalog No. 502913679
Encompass_Preferred
A secondary bile acid; a metabolite of deoxycholic acid in C. perfringens
Catalog No. 50-291-3679 Supplier Cayman Chemical Supplier No. 3522725 MG
A secondary bile acid; a metabolite of deoxycholic acid in C. perfringens