Learn More
Cayman Chemical 3-oxo DeoxycholIc AcId 50mg
SDP
Supplier:  Cayman Chemical 3522750 MG
Encompass_Preferred
A secondary bile acid; a metabolite of deoxycholic acid in C. perfringens
Catalog No. 50-291-3506