Learn More
Cayman Chemical 3-oxo Stearic Acid, Purity: ≥90%, CAS: 16694-29-4, 1 mg
SDP
Catalog No. 502971029
Encompass_Preferred
An intermediate in the dissociated (or type II) fatty acid biosynthesis that occurs in bacteria; generated following the reaction of hexadecenoyl-ACP with malonyl-ACP by the enzyme β-ketoacyl-ACP synthase II, which produces 3-oxo octadecenoyl-ACP
Catalog No. 50-297-1029 Supplier Cayman Chemical Supplier No. 148071 MG
An intermediate in the dissociated (or type II) fatty acid biosynthesis that occurs in bacteria; generated following the reaction of hexadecenoyl-ACP with malonyl-ACP by the enzyme β-ketoacyl-ACP synthase II, which produces 3-oxo octadecenoyl-ACP