Learn More
Cayman Chemical 3-oxo StearIc AcId 500ug
SDP
Catalog No. 502971941
Encompass_Preferred
An intermediate in the dissociated (or type II) fatty acid biosynthesis that occurs in bacteria; generated following the reaction of hexadecenoyl-ACP with malonyl-ACP by the enzyme β-ketoacyl-ACP synthase II, which produces 3-oxo octadecenoyl-ACP
Catalog No. 50-297-1941 Supplier Cayman Chemical Supplier No. 14807500 UG
An intermediate in the dissociated (or type II) fatty acid biosynthesis that occurs in bacteria; generated following the reaction of hexadecenoyl-ACP with malonyl-ACP by the enzyme β-ketoacyl-ACP synthase II, which produces 3-oxo octadecenoyl-ACP