Learn More
Cayman Chemical 3-Oxobetulin, Purity: ≥95%, CAS: 7020-34-0, 10 mg
SDP
Catalog No. 502947620
Encompass_Preferred
A pentacyclic triterpenoid; reduces HIV-1 replication in MT-2 cells (IC50 = 4.1 µM); inhibits cadmium-induced cytotoxicity in HepG2 cells in a concentration-dependent manner
Catalog No. 50-294-7620 Supplier Cayman Chemical Supplier No. 3306610 MG
A pentacyclic triterpenoid; reduces HIV-1 replication in MT-2 cells (IC50 = 4.1 µM); inhibits cadmium-induced cytotoxicity in HepG2 cells in a concentration-dependent manner