Learn More
Cayman Chemical 3-Oxobetulin Acetate, Purity: ≥95%, CAS: 136587-07-0, 5 mg
SDP
Catalog No. 502950731
Encompass_Preferred
A derivative of betulin; inhibits the growth of P388 cells (EC50 = 0.12 µg/ml), as well as MCF-7, SF-268, H460, and KM20L2 cells (GI50s = 8, 10.6, 5.2, and 12.7 µg/ml, respectively); inhibits replication of X4 tropic recombinant HIV (NL4.3-Ren) in MT-2 lymphoblastoid cells (IC50 = 13.4 µM); active against L. donovani amastigotes at 50 µM
Catalog No. 50-295-0731 Supplier Cayman Chemical Supplier No. 330675 MG
A derivative of betulin; inhibits the growth of P388 cells (EC50 = 0.12 µg/ml), as well as MCF-7, SF-268, H460, and KM20L2 cells (GI50s = 8, 10.6, 5.2, and 12.7 µg/ml, respectively); inhibits replication of X4 tropic recombinant HIV (NL4.3-Ren) in MT-2 lymphoblastoid cells (IC50 = 13.4 µM); active against L. donovani amastigotes at 50 µM