Learn More
TARGETMOL CHEMICALS INC 3-Oxocholic acid 10MG
SDP
Catalog No. 502878253
Encompass_Preferred
Also available in 1 mL, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 3-Oxocholic acid(3-Ketocholic acid) is the metabolite of bile acid and the main product of bile degradation by Clostridium perfringens in the intestine.3-Oxocholic acid is a steroid acid mainly found in the bile of mammals. Purity 99.63%
Catalog No. 50-287-8253 Supplier TARGETMOL CHEMICALS INC Supplier No. T1912410MG
Add to Cart
Add to Cart
Also available in 1 mL, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 3-Oxocholic acid(3-Ketocholic acid) is the metabolite of bile acid and the main product of bile degradation by Clostridium perfringens in the intestine.3-Oxocholic acid is a steroid acid mainly found in the bile of mammals. Purity 99.63%