Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Oxocholic acid | 2304-89-4 | 1 ML
SDP

Catalog No. 5000417653
Encompass_Preferred

3-Oxocholic acid is an oxo-bile acid metabolite and a major degradation product from cholic by C. perfringens in the intestine. It is a steroid acid found predominantly in the bile of mammals. This product is supplied as a 10 mM solution in DMSO, suitable for research applications.

  • Molecular weight: 406.56
  • Molecular formula: C24H38O5
  • Appearance: White to off-white solid
  • Classified as a steroid
  • Initial source: Endogenous metabolite
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (245.97 mM); requires sonication
  • For research use only

Catalog No. 50-004-17653 Supplier Medchemexpress LLC Supplier No. HYN738710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Oxocholic acid is an oxo-bile acid metabolite and a major degradation product from cholic by C. perfringens in the intestine. It is a steroid acid found predominantly in the bile of mammals. This product is supplied as a 10 mM solution in DMSO, suitable for research applications.

  • Molecular weight: 406.56
  • Molecular formula: C24H38O5
  • Appearance: White to off-white solid
  • Classified as a steroid
  • Initial source: Endogenous metabolite
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 100 mg/mL (245.97 mM); requires sonication
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.