Learn More
Cayman Chemical 3-Oxocholic Acid, Purity: ≥95%, CAS: 2304-89-4, 50 mg
SDP
Catalog No. 502940130
Encompass_Preferred
A secondary bile acid; a metabolite of cholic acid in C. perfringens; serum levels increase following ileal transposition surgery in rats
Catalog No. 50-294-0130 Supplier Cayman Chemical Supplier No. 2840350 MG
A secondary bile acid; a metabolite of cholic acid in C. perfringens; serum levels increase following ileal transposition surgery in rats