Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Oxopentanedioic acid | 542-05-2 | MFCD00002711 | 146.10 | 50 G
SDP

Catalog No. 5000446740
Encompass_Preferred

3-Oxopentanedioic acid is a dicarboxylic acid that is easily brominated and serves as an intermediate in organic chemical synthesis. This aliphatic β-dicarbonyl acid can also be utilized as a model compound to study the formation mechanism of brominated trihalomethanes during the chlorination, bromination, and ozonation of saline water.

  • Dicarboxylic acid, easily brominated
  • Intermediate in organic chemical synthesis
  • Model compound for studying brominated trihalomethane formation in saline water
  • Solid, white to off-white appearance
  • Store at -20°C, sealed, away from moisture
  • Soluble in H₂O (100 mg/mL) and DMSO (100 mg/mL); solutions are unstable

Catalog No. 50-004-46740 Supplier Medchemexpress LLC Supplier No. HYW00775250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Oxopentanedioic acid is a dicarboxylic acid that is easily brominated and serves as an intermediate in organic chemical synthesis. This aliphatic β-dicarbonyl acid can also be utilized as a model compound to study the formation mechanism of brominated trihalomethanes during the chlorination, bromination, and ozonation of saline water.

  • Dicarboxylic acid, easily brominated
  • Intermediate in organic chemical synthesis
  • Model compound for studying brominated trihalomethane formation in saline water
  • Solid, white to off-white appearance
  • Store at -20°C, sealed, away from moisture
  • Soluble in H₂O (100 mg/mL) and DMSO (100 mg/mL); solutions are unstable

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.