Learn More
MSC Dow Corning 4097353 3.04 oz Tube White RTV Silicone Joint Sealant -85 to 500°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 730
Catalog No. 501149218
Encompass
Dow Corning 4097353 3.04 oz Tube White RTV Silicone Joint Sealant -85 to 500°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 730
Catalog No. 50-114-9218 Supplier MSC Supplier No. 00231605
Dow Corning 4097353 3.04 oz Tube White RTV Silicone Joint Sealant -85 to 500°F Operating Temp, 14 min Tack Free Dry Time, 24 hr Full Cure Time, Series 730