Learn More
MSC Super Lube Silicone Dielectric Vacuum Grease, NSF H1, 3 oz
Catalog No. 501112926
Encompass
Super Lube® Silicone Dielectric and Vacuum Grease is a specially formulated non-curing silicone compound that performs well in both electrical and vacuum applications. It seals, protects, and insulates electrical components and connectors, providing a waterproof barrier against moisture and contaminants. It is an NSF H1 registered Food Grade lubricant.
  • High dielectric strength
  • Food grade and clean, Kosher certified
  • Seals out moisture to prevent rust and protect electrical components, connectors, O-rings, and rubber components
  • Prevents electrical arcing by filling microscopic air gaps between electrical components
  • Resistant to thermal degradation, water, and chemicals
  • Wide temperature range from -40°C to 260°C
  • Compatible with most rubber and plastic compounds
  • Low volatility and outgassing characteristics
Catalog No. 50-111-2926 Supplier MSC Supplier No. 02105658
Super Lube® Silicone Dielectric and Vacuum Grease is a specially formulated non-curing silicone compound that performs well in both electrical and vacuum applications. It seals, protects, and insulates electrical components and connectors, providing a waterproof barrier against moisture and contaminants. It is an NSF H1 registered Food Grade lubricant.
  • High dielectric strength
  • Food grade and clean, Kosher certified
  • Seals out moisture to prevent rust and protect electrical components, connectors, O-rings, and rubber components
  • Prevents electrical arcing by filling microscopic air gaps between electrical components
  • Resistant to thermal degradation, water, and chemicals
  • Wide temperature range from -40°C to 260°C
  • Compatible with most rubber and plastic compounds
  • Low volatility and outgassing characteristics