Learn More
MSC Super Lube Grease, Synthetic Syncolon, NLGI 2, 3 oz
Catalog No. 501112908
Encompass
This general purpose grease is a synthetic lubricant with Syncolon, ideal for a wide range of applications. It is NSF H1 certified for incidental food contact and Kosher Certified, offering excellent water resistance and rust inhibiting properties. The grease provides good mechanical stability across various temperatures and will not evaporate, form gummy deposits, melt, or separate.
  • NSF H1 certified for incidental food contact
  • Kosher certified
  • Excellent water resistance and rust inhibiting properties
  • Wide temperature range, effective in low and high-temperature conditions
  • Good mechanical stability
  • Does not evaporate or form gummy deposits
  • Will not melt or separate
  • Translucent color
  • NLGI grade 2
  • Synthetic base oil with PTFE thickener
  • VOC compliant
Catalog No. 50-111-2908 Supplier MSC Supplier No. 00257006
This general purpose grease is a synthetic lubricant with Syncolon, ideal for a wide range of applications. It is NSF H1 certified for incidental food contact and Kosher Certified, offering excellent water resistance and rust inhibiting properties. The grease provides good mechanical stability across various temperatures and will not evaporate, form gummy deposits, melt, or separate.
  • NSF H1 certified for incidental food contact
  • Kosher certified
  • Excellent water resistance and rust inhibiting properties
  • Wide temperature range, effective in low and high-temperature conditions
  • Good mechanical stability
  • Does not evaporate or form gummy deposits
  • Will not melt or separate
  • Translucent color
  • NLGI grade 2
  • Synthetic base oil with PTFE thickener
  • VOC compliant