Learn More
Medchemexpress LLC 3-Pentadecylphenol 25g
SDP
Catalog No. 5000204871
Encompass_Preferred
3-PentadecylphenolIt is an organic compound belonging to the phenolic class It is attached to the phenolic ring by 15Carbon pentadecyl chain composition with a touch of sweetness 3-Pentadecylphenol has various applications in the chemical and pharmaceutical industries especially as an intermediate in the synthesis of various chemicals and pharmaceuticals In addition it can be used as an antioxidant for plastics and rubber
Catalog No. 50-002-04871 Supplier Medchemexpress LLC Supplier No. 25GHYW127489
3-PentadecylphenolIt is an organic compound belonging to the phenolic class It is attached to the phenolic ring by 15Carbon pentadecyl chain composition with a touch of sweetness 3-Pentadecylphenol has various applications in the chemical and pharmaceutical industries especially as an intermediate in the synthesis of various chemicals and pharmaceuticals In addition it can be used as an antioxidant for plastics and rubber