Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Pentafluorophenyl)pentafluoro-1-propene 98.0+%, TCI America™
SDP

Catalog No. P13435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13435G Supplier TCI America Supplier No. P13435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67899-41-6
Molecular Formula C9F10
Molecular Weight (g/mol) 298.083
MDL Number MFCD00153225
InChI Key WRHBYJDZKRNITP-UHFFFAOYSA-N
Synonym perfluoro allylbenzene, perfluoroallylbenzene, 3-pentafluorophenyl pentafluoro-1-propene, 3-pentafluorophenyl pentafluoroprop-1-ene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-enyl benzene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-en-1-yl benzene, 1,2,3,4,5-pentafluoro-6-pentafluoroprop-2-en-1-yl benzene, benzene,1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoro-2-propen-1-yl, acmc-1b8oa, 1-pentafluoroallyl pentafluorobenzene
PubChem CID 2775989
IUPAC Name 1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-enyl)benzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(C(=C(F)F)F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 298.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Pentafluorophenyl)pentafluoro-1-propene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.