Learn More
Medchemexpress LLC 3-pentanol | 584-02-1 | 99.8% | 88.15 g/mol | C5H12O | 25 G
SDP
Catalog No. 5000209022
Encompass_Preferred
3-pentanol (pentan-3-ol) is a plant-derived organic compound and a component of insect pheromones supplied for research use. It has been reported to activate salicylic acid (SA) and jasmonic acid (JA) signaling pathways, triggering plant immune responses against microbial pathogens and pests.
  • Research-grade compound for laboratory studies
  • Reported activator of SA and JA signaling pathways
  • High purity (99.8%) suitable for analytical and biological assays
  • Available in multiple pack sizes, including 25 G
  • Storage recommendations: pure form at -20°C for long-term or 4°C for shorter-term storage
Catalog No. 50-002-09022 Supplier Medchemexpress LLC Supplier No. 25GHYW087988
3-pentanol (pentan-3-ol) is a plant-derived organic compound and a component of insect pheromones supplied for research use. It has been reported to activate salicylic acid (SA) and jasmonic acid (JA) signaling pathways, triggering plant immune responses against microbial pathogens and pests.
  • Research-grade compound for laboratory studies
  • Reported activator of SA and JA signaling pathways
  • High purity (99.8%) suitable for analytical and biological assays
  • Available in multiple pack sizes, including 25 G
  • Storage recommendations: pure form at -20°C for long-term or 4°C for shorter-term storage