Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pentanol | 584-02-1 | MFCD00004574 | 99.8% | 88.15 g/mol | C5H12O | 1 KG
SDP

Catalog No. 5000208421
Encompass_Preferred

3-pentanol (pentan-3-ol) is a secondary alcohol commonly used as a research reagent and chemical intermediate. It is used in studies of plant signaling and insect pheromones and is supplied as a high-purity liquid for laboratory use.

  • High purity: 99.76%.
  • CAS number: 584-02-1.
  • Chemical formula C5H12O, molecular weight 88.15 g/mol.
  • Physical state: colorless to light yellow liquid; density ~0.8 g/cm3.
  • Available in multiple pack sizes including 1 kg.
  • Includes analytical documents: data sheet, COA, SDS, GC, H-NMR.
  • Storage: store cold; pure form stable at -20°C for years.

Catalog No. 50-002-08421 Supplier Medchemexpress LLC Supplier No. HYW0879881KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-pentanol (pentan-3-ol) is a secondary alcohol commonly used as a research reagent and chemical intermediate. It is used in studies of plant signaling and insect pheromones and is supplied as a high-purity liquid for laboratory use.

  • High purity: 99.76%.
  • CAS number: 584-02-1.
  • Chemical formula C5H12O, molecular weight 88.15 g/mol.
  • Physical state: colorless to light yellow liquid; density ~0.8 g/cm3.
  • Available in multiple pack sizes including 1 kg.
  • Includes analytical documents: data sheet, COA, SDS, GC, H-NMR.
  • Storage: store cold; pure form stable at -20°C for years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.