Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pentanol | 584-02-1 | MFCD00004574 | 99.8% | 88.15 g/mol | C5H12O | 1 ML
SDP

Catalog No. 5000208416
Encompass_Preferred

3-pentanol is a secondary alcohol (chemical formula C5H12O) supplied as a 10 mM solution in a 1 mL vial for laboratory research use. CAS number 584-02-1; purity by gas chromatography 99.76% (reported).

  • Secondary alcohol commonly used as a reagent and research chemical
  • Cas number 584-02-1
  • Molecular weight 88.15 g/mol
  • Concentration 10 mM in a 1 mL solution
  • Purity (GC) 99.8%
  • Physical state colorless liquid

Catalog No. 50-002-08416 Supplier Medchemexpress LLC Supplier No. HYW08798810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-pentanol is a secondary alcohol (chemical formula C5H12O) supplied as a 10 mM solution in a 1 mL vial for laboratory research use. CAS number 584-02-1; purity by gas chromatography 99.76% (reported).

  • Secondary alcohol commonly used as a reagent and research chemical
  • Cas number 584-02-1
  • Molecular weight 88.15 g/mol
  • Concentration 10 mM in a 1 mL solution
  • Purity (GC) 99.8%
  • Physical state colorless liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.