Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pentanol | 584-02-1 | MFCD00004574 | 99.8% | 88.15 g/mol | C5H12O | 50 G
SDP

Catalog No. 5000208431
Encompass_Preferred

3-pentanol (pentan-3-ol) is a secondary alcohol used as a research reagent and reference standard. It is supplied for research use and is commonly used in studies of plant signaling, insect pheromones, and as a GC reference standard.

  • high purity suitable for analytical use
  • secondary alcohol with molecular formula C5H12O
  • molecular weight 88.15 g/mol
  • CAS number 584-02-1
  • available in multiple pack sizes including 50 G

Catalog No. 50-002-08431 Supplier Medchemexpress LLC Supplier No. HYW08798850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-pentanol (pentan-3-ol) is a secondary alcohol used as a research reagent and reference standard. It is supplied for research use and is commonly used in studies of plant signaling, insect pheromones, and as a GC reference standard.

  • high purity suitable for analytical use
  • secondary alcohol with molecular formula C5H12O
  • molecular weight 88.15 g/mol
  • CAS number 584-02-1
  • available in multiple pack sizes including 50 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.