Learn More
Medchemexpress LLC 3-pentanol-d5 | 144032-75-7 | 93.18 | C5H7D5O | 10 MG
SDP
Catalog No. 5000208279
Encompass_Preferred
3-Pentanol-d5 is the deuterium-labeled analogue of 3-pentanol used as an isotopic internal standard and reference material for analytical chemistry and mass spectrometry applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Liquid form suitable for GC-MS and LC-MS standard preparation.
  • Offered in small mg-scale pack sizes for standards and calibration.
  • Molecular weight 93.18 and chemical formula C5H7D5O.
Catalog No. 50-002-08279 Supplier Medchemexpress LLC Supplier No. HYW087988S10MG
3-Pentanol-d5 is the deuterium-labeled analogue of 3-pentanol used as an isotopic internal standard and reference material for analytical chemistry and mass spectrometry applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Liquid form suitable for GC-MS and LC-MS standard preparation.
  • Offered in small mg-scale pack sizes for standards and calibration.
  • Molecular weight 93.18 and chemical formula C5H7D5O.