Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pentanol-d5 | 144032-75-7 | 93.18 | C5H7D5O | 50 MG
SDP

Catalog No. 5000208331
Encompass_Preferred

3-Pentanol-d5 is the deuterium-labeled form of 3-pentanol used as an isotopic tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is described as a plant-produced organic compound and a component of insect sex pheromones; stable heavy isotopes may affect pharmacokinetic and metabolic profiles.

  • Deuterium-labeled isotopic standard for analytical applications.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • Available in small pack sizes for analytical use, including 50 mg.
  • CAS number 144032-75-7; molecular formula C5H7D5O; molecular weight 93.18.
  • Purity documentation is available on the product page.

Catalog No. 50-002-08331 Supplier Medchemexpress LLC Supplier No. HYW087988S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Pentanol-d5 is the deuterium-labeled form of 3-pentanol used as an isotopic tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is described as a plant-produced organic compound and a component of insect sex pheromones; stable heavy isotopes may affect pharmacokinetic and metabolic profiles.

  • Deuterium-labeled isotopic standard for analytical applications.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • Available in small pack sizes for analytical use, including 50 mg.
  • CAS number 144032-75-7; molecular formula C5H7D5O; molecular weight 93.18.
  • Purity documentation is available on the product page.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.