Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pentanol-d5 | 144032-75-7 | 93.18 g/mol | C5H7D5O | 100 MG
SDP

Catalog No. 5000208333
Encompass_Preferred

3-pentanol-d5 is the deuterium-labeled form of 3-pentanol used as a stable isotope tracer and internal standard for analytical techniques such as NMR, GC-MS, and LC-MS. It is supplied as a liquid deuterated compound for research and quantitative analysis.

  • Deuterium-labeled internal standard for quantitative analysis
  • Molecular formula: C5H7D5O
  • Molecular weight: 93.18 g/mol
  • Appearance: liquid
  • Available pack sizes: 10 mg, 25 mg, 50 mg, 100 mg, 200 mg, 500 mg
  • Recommended storage: pure form -20°C (stable ~3 years); in solvent -80°C (up to 6 months), -20°C (up to 1 month)

Catalog No. 50-002-08333 Supplier Medchemexpress LLC Supplier No. HYW087988S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-pentanol-d5 is the deuterium-labeled form of 3-pentanol used as a stable isotope tracer and internal standard for analytical techniques such as NMR, GC-MS, and LC-MS. It is supplied as a liquid deuterated compound for research and quantitative analysis.

  • Deuterium-labeled internal standard for quantitative analysis
  • Molecular formula: C5H7D5O
  • Molecular weight: 93.18 g/mol
  • Appearance: liquid
  • Available pack sizes: 10 mg, 25 mg, 50 mg, 100 mg, 200 mg, 500 mg
  • Recommended storage: pure form -20°C (stable ~3 years); in solvent -80°C (up to 6 months), -20°C (up to 1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.