Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-pentanol-d5 | 144032-75-7 | 93.18 g/mol | C5H7D5O | 25 MG
SDP

Catalog No. 5000209643
Encompass_Preferred

3-pentanol-d5 is the deuterium-labeled analog of 3-pentanol used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It supports analytical workflows that require isotopic labeling for tracing and accurate quantification.

  • Deuterium-labeled 3-pentanol for use as a tracer and internal standard.
  • Intended for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Molecular formula: C5H7D5O.
  • Molecular weight: 93.18 g/mol.
  • CAS number: 144032-75-7.
  • Available pack size: 25 mg.
  • Storage: pure form -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-09643 Supplier Medchemexpress LLC Supplier No. 25MGHYW087988S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-pentanol-d5 is the deuterium-labeled analog of 3-pentanol used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It supports analytical workflows that require isotopic labeling for tracing and accurate quantification.

  • Deuterium-labeled 3-pentanol for use as a tracer and internal standard.
  • Intended for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Molecular formula: C5H7D5O.
  • Molecular weight: 93.18 g/mol.
  • CAS number: 144032-75-7.
  • Available pack size: 25 mg.
  • Storage: pure form -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.