Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Perfluorobutyryl)-(+)-Camphor

Catalog No. 1959365G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-593-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 19-593-65G Supplier Sigma Aldrich Supplier No. 1959365G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(Perfluorobutyryl)-(+)-camphor can be used:As a ligand in the synthesis of chiral europium complex for optoelectronic and photonic applications.As a ligand in the preparation of sodium tetrakis(3-heptafluorobutylryl-(+)-camphorato) Ln(III) complexes applicable as chemical sensors and NMR shift reagents.

Specifications

CAS 51800-98-7
Density 1.172 g/mL at 25°C
Boiling Point 100°C to 101°C (16 mmHg, lit.)
Molecular Formula C12H15F3O2
Refractive Index n20/D 1.421
Linear Formula C12H15F3O2
MDL Number MFCD00003746
Quantity 5 g
Synonym (+)-3-(Heptafluorobutyryl)-(+)-camphor; 3-(Perfluorobutyryl)-D-camphor
Molecular Weight (g/mol) 248.24 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.