Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Phenoxybenzaldehyde

Catalog No. 19175225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-175-225G 25 g
19-175-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-175-225G Supplier Sigma Aldrich Supplier No. 19175225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Enantioselective autoinduction during asymmetric hydrocyanation of 3-phenoxybenzaldehyde catalyzed by cyclo[(r)-phenylalanyl-(r)-histidyl] has been investigated
  • It undergoes hydrogenation catalyzed by au/pt bimetallic core/shell nanoparticles to yield 3-phenoxyphenyl methanol

Specifications

CAS 39515-51-0
Density 1.147 g/mL (at 25°C (literature))
Boiling Point 169°C to 169.5°C (11 mmHg)
Molecular Formula C13H10O2
Refractive Index n20/D 1.595 (literature)
Linear Formula C6H5OC6H4CHO
MDL Number MFCD00003353
Quantity 25 g
Molecular Weight (g/mol) 198.22
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.