Learn More
Medchemexpress LLC 3-phenoxybenzoic acid | 3739-38-6 | MFCD00002498 | 98.2% | 214.22 g/mol | C13H10O3 | 250 MG
SDP
Catalog No. 5000303726
Encompass_Preferred
3-Phenoxybenzoic acid analytical standard intended for research and analytical applications. It serves as a reference material for calibration, method development, and quantitative testing of environmental and biological samples. The compound is a metabolite of pyrethroid insecticides and is supplied as a solid with reported purity of approximately 98.2% and a molecular weight of 214.22 g/mol.
  • Provides high-purity reference material for analytical calibration.
  • Suitable for environmental and biological quantitative analysis.
  • Available in multiple pack sizes including 250 mg for flexible use.
  • Stable solid form for routine storage and handling.
  • Supports method development and validation workflows.
Catalog No. 50-003-03726 Supplier Medchemexpress LLC Supplier No. 250MGHYW014225R
3-Phenoxybenzoic acid analytical standard intended for research and analytical applications. It serves as a reference material for calibration, method development, and quantitative testing of environmental and biological samples. The compound is a metabolite of pyrethroid insecticides and is supplied as a solid with reported purity of approximately 98.2% and a molecular weight of 214.22 g/mol.
  • Provides high-purity reference material for analytical calibration.
  • Suitable for environmental and biological quantitative analysis.
  • Available in multiple pack sizes including 250 mg for flexible use.
  • Stable solid form for routine storage and handling.
  • Supports method development and validation workflows.