Learn More
Medchemexpress LLC 3-phenoxypropyl bromide | 588-63-6 | MFCD00000256 | 97.1% | 215.09 g/mol | C9H11BrO | 50 G
SDP
Catalog No. 5000300918
Encompass_Preferred
3-Phenoxypropyl bromide (CAS 588-63-6) is an organic reagent used in life-science and synthetic chemistry as an alkylating agent and intermediate for preparing phenoxypropyl derivatives. It is a colorless to light yellow liquid with formula C9H11BrO and molecular weight 215.09 g/mol.
  • Functions as an alkylating agent and synthetic intermediate.
  • Colorless to light yellow liquid for easy handling.
  • Manufacturer-listed purity around 97.1% for research use.
  • Available in multiple pack sizes (10 g-100 g) for scale flexibility.
  • Suitable for organic synthesis and life-science research applications.
Catalog No. 50-003-00918 Supplier Medchemexpress LLC Supplier No. HYW01962750G
3-Phenoxypropyl bromide (CAS 588-63-6) is an organic reagent used in life-science and synthetic chemistry as an alkylating agent and intermediate for preparing phenoxypropyl derivatives. It is a colorless to light yellow liquid with formula C9H11BrO and molecular weight 215.09 g/mol.
  • Functions as an alkylating agent and synthetic intermediate.
  • Colorless to light yellow liquid for easy handling.
  • Manufacturer-listed purity around 97.1% for research use.
  • Available in multiple pack sizes (10 g-100 g) for scale flexibility.
  • Suitable for organic synthesis and life-science research applications.