Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenyl-1-propyne, 97%, stabilized, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
10147-11-2
C9H8
116.16
MFCD00134431
NGKSKVYWPINGLI-UHFFFAOYSA-N
3-phenyl-1-propyne,2-propynyl benzene,benzylacetylene,prop-2-yn-1-ylbenzene,benzene, 2-propynyl,propargylbenzol,propargylbenzene,benzyl acetylene,3-phenyl-propyne,3-phenyl-1-propyn
575753
prop-2-ynylbenzene
C#CCC1=CC=CC=C1
10147-11-2
C9H8
116.16
MFCD00134431
NGKSKVYWPINGLI-UHFFFAOYSA-N
3-phenyl-1-propyne,2-propynyl benzene,benzylacetylene,prop-2-yn-1-ylbenzene,benzene, 2-propynyl,propargylbenzol,propargylbenzene,benzyl acetylene,3-phenyl-propyne,3-phenyl-1-propyn
575753
prop-2-ynylbenzene
C#CCC1=CC=CC=C1
Specific Gravity 0.934
Refractive Index 1.5260 to 1.5290
Density 0.9340g/mL
Boiling Point 75.0°C (20.0 mmHg)
Packaging Glass bottle
Stabilizer 250ppm BHT
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 25g
Flash Point 52°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 116.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Phenyl-1-propyne, Stabilized
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning