Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) Dichloride Tetrahydrofuran Adduct 90.0+%, TCI America™
SDP

Catalog No. P1688100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1688100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1688100MG Supplier TCI America Supplier No. P1688100MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 250220-36-1
Molecular Formula C51H76Cl2P2Ru
Molecular Weight (g/mol) 923.087
MDL Number MFCD07369034
InChI Key UZAFZWUWHHFWOK-UHFFFAOYSA-L
PubChem CID 11029366
IUPAC Name dichloro-(3-phenylinden-1-ylidene)ruthenium;tricyclohexylphosphane
SMILES C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.C1=CC=C(C=C1)C2=CC(=[Ru](Cl)Cl)C3=CC=CC=C32

Specifications

Quantity 100 mg
Formula Weight 923.09
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) Dichloride Tetrahydrofuran Add
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.