Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenylazo-2,6-diaminopyridine Monohydrochloride 98.0+%, TCI America™
SDP

Catalog No. B002225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B002225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B002225G Supplier TCI America Supplier No. B002225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity chemical for research use, with a minimum purity of 98.0%.

Chemical Identifiers

CAS 136-40-3
Molecular Formula C11H12ClN5
Molecular Weight (g/mol) 249.702
MDL Number MFCD00035347
InChI Key QQBPIHBUCMDKFG-UHFFFAOYSA-N
Synonym phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine
PubChem CID 8691
ChEBI CHEBI:71419
IUPAC Name 3-phenyldiazenylpyridine-2,6-diamine;hydrochloride
SMILES C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl

Specifications

Melting Point 238°C
Color Purple-Red
Quantity 25 g
Formula Weight 249.70
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylazo-2,6-diaminopyridine Monohydrochloride
RTECSNumber US7875000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.